siRNA Details
| Gene Symbol: Amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031011.3 | NP_112273.3 | 3 | 633 | ||
| GenBank mRNA | AY724509.1 | - | 3 | 316 | ||
| BC061532.1 | - | 3 | 636 | 6921099 | ||
| FM031711.1 | - | 3 | 280 | |||
| FQ232080.1 | - | 3 | 453 | |||
| FQ234346.1 | - | 3 | 456 | |||
| M34464.1 | - | 3 | 579 | |||

Amd1
Genomic Map