siRNA Details
| Gene Symbol: Amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031011.3 | NP_112273.3 | 6, 7 | 1030 | ||
| GenBank mRNA | AY724509.1 | - | 6, 7 | 713 | ||
| BC061532.1 | - | 6, 7 | 1033 | 6921099 | ||
| FM123317.1 | - | 1, 2 | 227 | |||
| FQ232080.1 | - | 6, 7 | 850 | |||
| FQ234346.1 | - | 6, 7 | 853 | |||
| FQ234398.1 | - | 1, 2 | 1199 | |||
| M34464.1 | - | 6, 7 | 976 | |||

Amd1
Genomic Map