siRNA Details
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031020.2 | NP_112282.2 | 11 | 1304 | ||
XM_006256142.2 | XP_006256204.1 | 11 | 1016 | |||
GenBank mRNA | AB000783.1 | - | 11 | 1303 | ||
AF346293.1 | - | 11 | 1001 | |||
BC092193.1 | - | 11 | 1304 | 7313217 | ||
U91847.1 | - | 11 | 1001 |
