siRNA Details
Gene Symbol: Gna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031033.1 | NP_112295.1 | 7 | 924 | ||
GenBank mRNA | AF239674.1 | - | 7 | 924 | ||
L37293.1 | - | 2 | 195 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031033.1 | NP_112295.1 | 7 | 924 | ||
GenBank mRNA | AF239674.1 | - | 7 | 924 | ||
L37293.1 | - | 2 | 195 |