siRNA Details
Gene Symbol: Gna12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031034.2 | NP_112296.1 | 4 | 963 | ||
GenBank mRNA | D85760.1 | - | 4 | 997 | ||
DQ120479.1 | - | 4 | 932 | |||
DQ120480.1 | - | 4 | 932 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gna12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031034.2 | NP_112296.1 | 4 | 963 | ||
GenBank mRNA | D85760.1 | - | 4 | 997 | ||
DQ120479.1 | - | 4 | 932 | |||
DQ120480.1 | - | 4 | 932 |