siRNA Details
| Gene Symbol: Gnai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031035.3 | NP_112297.1 | 7 | 1010 | ||
| XM_006243858.1 | XP_006243920.1 | 7 | 1040 | |||
| GenBank mRNA | AY899210.1 | - | 7 | 858 | ||
| BC078806.1 | - | 7 | 962 | 7121183 | ||
| DQ120471.1 | - | 7 | 919 | |||
| DQ120472.1 | - | 7 | 919 | |||
| M12672.1 | - | 7 | 906 | |||
| M17528.1 | - | 7 | 1008 | |||

Gnai2
Genomic Map