siRNA Details
| Gene Symbol: Gnai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031035.3 | NP_112297.1 | 8 | 1064 | ||
| XM_006243858.1 | XP_006243920.1 | 8 | 1094 | |||
| GenBank mRNA | BC078806.1 | - | 8 | 1016 | 7121183 | |
| DQ120471.1 | - | 8 | 973 | |||
| DQ120472.1 | - | 8 | 973 | |||
| M12672.1 | - | 8 | 960 | |||
| M17528.1 | - | 8 | 1062 | |||

Gnai2
Genomic Map