siRNA Details
| Gene Symbol: Itpka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031045.2 | NP_112307.2 | 6 | 1192 | ||
| GenBank mRNA | BQ204027.1 | - | 2 | 636 | ||
| M29787.1 | - | 6 | 1192 | |||
| X56917.1 | - | 6 | 1183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpka| Gene Symbol: Itpka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031045.2 | NP_112307.2 | 6 | 1192 | ||
| GenBank mRNA | BQ204027.1 | - | 2 | 636 | ||
| M29787.1 | - | 6 | 1192 | |||
| X56917.1 | - | 6 | 1183 | |||