siRNA Details
| Gene Symbol: Snrpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031117.2 | NP_112379.1 | 5 | 676 | ||
| GenBank mRNA | BC087671.1 | - | 1 | 80 | 7316987 | |
| FM122235.1 | - | 4 | 472 | |||
| M29293.1 | - | 5 | 676 | |||
| Gene Symbol: Snurf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270712.1 | NP_001257641.1 | 6 | 700 | ||
| NM_130738.2 | NP_570094.1 | 5 | 565 | |||

Snrpn
Genomic Map