siRNA Details
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031131.1 | NP_112393.1 | 3 | 772 | ||
XM_006250448.2 | XP_006250510.1 | 2 | 1598 | |||
GenBank mRNA | AF135598.1 | - | 2 | 382 | ||
AF153012.2 | - | 3 | 772 | |||
AF153013.1 | - | 2 | 688 | |||
AJ132718.1 | - | 2 | 396 |
