siRNA Details
| Gene Symbol: Klf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031135.2 | NP_112397.1 | 4 | 1436 | ||
| GenBank mRNA | BC097309.1 | - | 4 | 1436 | 7387595 | |
| U78875.1 | - | 4 | 1705 | |||
| U88630.1 | - | 3 | 1342 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf10| Gene Symbol: Klf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031135.2 | NP_112397.1 | 4 | 1436 | ||
| GenBank mRNA | BC097309.1 | - | 4 | 1436 | 7387595 | |
| U78875.1 | - | 4 | 1705 | |||
| U88630.1 | - | 3 | 1342 | |||