siRNA Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031139.1 | NP_112401.1 | 5, 6 | 1098 | ||
GenBank mRNA | AB035647.1 | - | 5, 6 | 587 | ||
AB035648.1 | - | 5, 6 | 506 | |||
AB035649.1 | - | 4, 5 | 386 | |||
AB035650.1 | - | 4, 5 | 540 | |||
AB035651.1 | - | 2, 3 | 220 | |||
AB047556.1 | - | 5, 6 | 1098 | |||
BC128735.1 | - | 5, 6 | 592 | 7190049 | ||
X90823.1 | - | 3, 4 | 422 |
