siRNA Details
| Gene Symbol: Usf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031139.1 | NP_112401.1 | 8 | 1303 | ||
| GenBank mRNA | AB035647.1 | - | 8 | 792 | ||
| AB035648.1 | - | 8 | 711 | |||
| AB035649.1 | - | 7 | 591 | |||
| AB035650.1 | - | 7 | 745 | |||
| AB035651.1 | - | 5 | 425 | |||
| AB047556.1 | - | 8 | 1303 | |||
| BC128735.1 | - | 8 | 797 | 7190049 | ||
| X90823.1 | - | 6 | 627 | |||

Usf2
Genomic Map