siRNA Details
| Gene Symbol: Usf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031139.1 | NP_112401.1 | 9 | 1354 | ||
| GenBank mRNA | AB035647.1 | - | 9 | 843 | ||
| AB035648.1 | - | 9 | 762 | |||
| AB035649.1 | - | 8 | 642 | |||
| AB035650.1 | - | 8 | 796 | |||
| AB035651.1 | - | 6 | 476 | |||
| AB047556.1 | - | 9 | 1354 | |||
| BC128735.1 | - | 9 | 848 | 7190049 | ||
| X90823.1 | - | 7 | 678 | |||

Usf2
Genomic Map