siRNA Details
| Gene Symbol: Vim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031140.1 | NP_112402.1 | 1 | 389 | ||
| GenBank mRNA | BC061847.1 | - | 1 | 429 | 5621065 | |
| FQ234991.1 | - | 1 | 143 | |||
| M84481.1 | - | 1 | 55 | |||
| X62952.1 | - | 1 | 389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vim| Gene Symbol: Vim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031140.1 | NP_112402.1 | 1 | 389 | ||
| GenBank mRNA | BC061847.1 | - | 1 | 429 | 5621065 | |
| FQ234991.1 | - | 1 | 143 | |||
| M84481.1 | - | 1 | 55 | |||
| X62952.1 | - | 1 | 389 | |||