siRNA Details
Gene Symbol: Actb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031144.3 | NP_112406.1 | 4 | 701 | ||
GenBank mRNA | BC063166.1 | - | 4 | 704 | 6920838 | |
EF156276.1 | - | 2 | 423 | |||
FQ227027.1 | - | 263 | ||||
FQ227201.1 | - | 60 | ||||
FQ228822.1 | - | 282 | ||||
FQ229293.1 | - | 62 | ||||
FQ231277.1 | - | 57 | ||||
FQ231503.1 | - | 4 | 703 | |||
FQ231691.1 | - | 51 | ||||
FQ232063.1 | - | 4 | 703 | |||
FQ232650.1 | - | 4 | 703 | |||
FQ232682.1 | - | 243 |
