siRNA Details
Gene Symbol: Lypla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031342.1 | NP_112632.1 | 7 | 414 | ||
GenBank mRNA | AB021645.1 | - | 7 | 414 | ||
BC070503.1 | - | 7 | 420 | 7096587 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lypla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031342.1 | NP_112632.1 | 7 | 414 | ||
GenBank mRNA | AB021645.1 | - | 7 | 414 | ||
BC070503.1 | - | 7 | 420 | 7096587 |