siRNA Details
| Gene Symbol: Tsc22d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031345.1 | NP_112635.1 | 1 | 306 | ||
| GenBank mRNA | AB025431.1 | - | 1 | 306 | ||
| BC061979.1 | - | 1 | 304 | 5598145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsc22d3| Gene Symbol: Tsc22d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031345.1 | NP_112635.1 | 1 | 306 | ||
| GenBank mRNA | AB025431.1 | - | 1 | 306 | ||
| BC061979.1 | - | 1 | 304 | 5598145 | ||