siRNA Details
| Gene Symbol: Tsc22d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031345.1 | NP_112635.1 | 2, 3 | 382 | ||
| XM_006234249.2 | XP_006234311.1 | 3, 4 | 649 | |||
| XM_006234250.2 | XP_006234312.1 | 3, 4 | 563 | |||
| XM_006234251.2 | XP_006234313.1 | 3, 4 | 598 | |||
| XM_008773446.1 | XP_008771668.1 | 2, 3 | 231 | |||
| XM_008773447.1 | XP_008771669.1 | 3, 4 | 502 | |||
| GenBank mRNA | AB025431.1 | - | 2, 3 | 382 | ||
| BC061979.1 | - | 2, 3 | 380 | 5598145 | ||

Tsc22d3
Genomic Map