siRNA Details
| Gene Symbol: Apba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031779.2 | NP_113967.1 | 3 | 1584 | ||
| XM_006231185.2 | XP_006231247.1 | 2 | 1717 | |||
| XM_008760312.1 | XP_008758534.1 | 2 | 1717 | |||
| GenBank mRNA | AF029105.1 | - | 1804 | |||

Apba1
Genomic Map