siRNA Details
| Gene Symbol: Pax4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031799.1 | NP_113987.1 | 3 | 364 | ||
| XM_006236231.2 | XP_006236293.1 | 4 | 6417 | |||
| XM_006236232.2 | XP_006236294.1 | 5 | 1413 | |||
| XM_006236233.2 | XP_006236295.1 | 5 | 855 | |||
| GenBank mRNA | AF053100.1 | - | 3 | 364 | ||
| AF053101.1 | - | 3 | 364 | |||
| AF053102.1 | - | 3 | 364 | |||
| AF053103.1 | - | 3 | 364 | |||
| AF198155.1 | - | 2 | 316 | |||
| AF198156.1 | - | 2 | 316 | |||

Pax4
Genomic Map