siRNA Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031801.1 | NP_113989.1 | 4 | 735 | ||
| XM_006230582.2 | XP_006230644.1 | 4 | 1603 | |||
| XM_006230583.2 | XP_006230645.1 | 4 | 1603 | |||
| XM_008760011.1 | XP_008758233.1 | 4 | 654 | |||
| XM_008760012.1 | XP_008758234.1 | 4 | 654 | |||
| XM_008760013.1 | XP_008758235.1 | 4 | 654 | |||
| XM_008760014.1 | XP_008758236.1 | 4 | 1619 | |||
| GenBank mRNA | AF055884.1 | - | 4 | 735 | ||
| U59659.1 | - | 4 | 484 | |||

Deaf1
Genomic Map