siRNA Details
| Gene Symbol: Tpbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031807.1 | NP_113995.1 | 2 | 1616 | ||
| GenBank mRNA | AF063939.1 | - | 2 | 1616 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpbg| Gene Symbol: Tpbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031807.1 | NP_113995.1 | 2 | 1616 | ||
| GenBank mRNA | AF063939.1 | - | 2 | 1616 | ||