siRNA Details
Gene Symbol: Clta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031974.1 | NP_114180.1 | 4, 5 | 611 | ||
XM_006238104.1 | XP_006238166.1 | 4, 5 | 1384 | |||
GenBank mRNA | FQ213034.1 | - | 4, 5 | 619 | ||
FQ213766.1 | - | 4, 5 | 612 | |||
M15882.1 | - | 4, 5 | 611 | |||
M19260.1 | - | 4, 5 | 611 |
