siRNA Details
| Gene Symbol: Gipc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053341.1 | NP_445793.1 | 3, 4 | 699 | ||
| XM_006255311.2 | XP_006255373.1 | 3, 4 | 2964 | |||
| XM_006255312.2 | XP_006255374.1 | 4, 5 | 1958 | |||
| XM_008772420.1 | XP_008770642.1 | 4, 5 | 1888 | |||
| GenBank mRNA | AF032120.1 | - | 3, 4 | 699 | ||
| AF089817.1 | - | 4, 5 | 717 | |||
| BC061964.1 | - | 2, 3 | 1507 | |||
| BC070505.1 | - | 4, 5 | 742 | 7097697 | ||

Gipc1
Genomic Map