siRNA Details
| Gene Symbol: Gipc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053341.1 | NP_445793.1 | 3 | 574 | ||
| XM_006255311.2 | XP_006255373.1 | 3 | 2839 | |||
| XM_006255312.2 | XP_006255374.1 | 4 | 1833 | |||
| XM_008772420.1 | XP_008770642.1 | 4 | 1763 | |||
| GenBank mRNA | AF032120.1 | - | 3 | 574 | ||
| AF089817.1 | - | 4 | 592 | |||
| BC070505.1 | - | 4 | 617 | 7097697 | ||

Gipc1
Genomic Map