siRNA Details
Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031985.1 | NP_114191.1 | 7, 8 | 699 | ||
XM_006247111.2 | XP_006247173.1 | 7, 8 | 836 | |||
XM_008768120.1 | XP_008766342.1 | 7, 8 | 1100 | |||
XM_008768121.1 | XP_008766343.1 | 8, 9 | 722 | |||
GenBank mRNA | BC089789.1 | - | 8, 9 | 769 | ||
M58340.1 | - | 7, 8 | 699 |
