siRNA Details
| Gene Symbol: Rps6kb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031985.1 | NP_114191.1 | 7 | 623 | ||
| XM_006247111.2 | XP_006247173.1 | 7 | 760 | |||
| XM_008768120.1 | XP_008766342.1 | 7 | 1024 | |||
| XM_008768121.1 | XP_008766343.1 | 8 | 646 | |||
| GenBank mRNA | BC089789.1 | - | 8 | 693 | ||
| M58340.1 | - | 7 | 623 | |||

Rps6kb1
Genomic Map