siRNA Details
| Gene Symbol: Slc1a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032065.1 | NP_114454.1 | 6 | 1108 | ||
| XM_008765182.1 | XP_008763404.1 | 6 | 1282 | |||
| GenBank mRNA | AY286330.1 | - | 6 | 1102 | ||
| BC098912.1 | - | 6 | 1132 | 7315936 | ||
| U89608.1 | - | 6 | 1108 | |||

Slc1a6
Genomic Map