siRNA Details
| Gene Symbol: Hsd17b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032066.1 | NP_114455.1 | 9, 10 | 700 | ||
| XM_006234599.2 | XP_006234661.1 | 9, 10 | 867 | |||
| GenBank mRNA | BC061543.1 | - | 9, 10 | 721 | 6888949 | |
| FQ213557.1 | - | 9, 10 | 736 | |||
| FQ214777.1 | - | 9, 10 | 765 | |||
| U81186.1 | - | 9, 10 | 700 | |||

Hsd17b12
Genomic Map