siRNA Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC127481.1 | - | 8, 9 | 674 | 8370668 | |
| FM118995.1 | - | 2, 3 | 240 | |||
| U90829.1 | - | 8, 9 | 671 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nae1| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC127481.1 | - | 8, 9 | 674 | 8370668 | |
| FM118995.1 | - | 2, 3 | 240 | |||
| U90829.1 | - | 8, 9 | 671 | |||