siRNA Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC127481.1 | - | 9 | 708 | 8370668 | |
| FM118995.1 | - | 3 | 274 | |||
| U90829.1 | - | 9 | 705 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nae1| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC127481.1 | - | 9 | 708 | 8370668 | |
| FM118995.1 | - | 3 | 274 | |||
| U90829.1 | - | 9 | 705 | |||