siRNA Details
| Gene Symbol: Cacng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053351.1 | NP_445803.1 | 3, 4 | 468 | ||
| GenBank mRNA | AF118818.1 | - | 3, 4 | 468 | ||
| AF361339.1 | - | 3, 4 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacng2| Gene Symbol: Cacng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053351.1 | NP_445803.1 | 3, 4 | 468 | ||
| GenBank mRNA | AF118818.1 | - | 3, 4 | 468 | ||
| AF361339.1 | - | 3, 4 | 435 | |||