siRNA Details
| Gene Symbol: Il12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053390.1 | NP_445842.1 | 2 | 135 | ||
| GenBank mRNA | AF177031.1 | - | 2 | 135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il12a| Gene Symbol: Il12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053390.1 | NP_445842.1 | 2 | 135 | ||
| GenBank mRNA | AF177031.1 | - | 2 | 135 | ||