siRNA Details
| Gene Symbol: Wnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053402.2 | NP_445854.1 | 3 | 340 | ||
| GenBank mRNA | AF188608.1 | - | 3 | 341 | ||
| BC098752.1 | - | 3 | 361 | 7369699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt4| Gene Symbol: Wnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053402.2 | NP_445854.1 | 3 | 340 | ||
| GenBank mRNA | AF188608.1 | - | 3 | 341 | ||
| BC098752.1 | - | 3 | 361 | 7369699 | ||