siRNA Details
| Gene Symbol: Tmed10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053467.1 | NP_445919.1 | 2 | 278 | ||
| GenBank mRNA | AJ004912.1 | - | 2 | 278 | ||
| FQ212056.1 | - | 2 | 312 | |||
| X97443.1 | - | 2 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmed10| Gene Symbol: Tmed10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053467.1 | NP_445919.1 | 2 | 278 | ||
| GenBank mRNA | AJ004912.1 | - | 2 | 278 | ||
| FQ212056.1 | - | 2 | 312 | |||
| X97443.1 | - | 2 | 230 | |||