siRNA Details
| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058208.1 | NP_478115.1 | 2 | 534 | ||
| GenBank mRNA | AF075382.1 | - | 2 | 534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs2| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058208.1 | NP_478115.1 | 2 | 534 | ||
| GenBank mRNA | AF075382.1 | - | 2 | 534 | ||