siRNA Details
| Gene Symbol: Bmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191856.2 | NP_001178785.1 | 2 | 967 | ||
| GenBank mRNA | AF100787.1 | - | 2 | 212 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp7| Gene Symbol: Bmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191856.2 | NP_001178785.1 | 2 | 967 | ||
| GenBank mRNA | AF100787.1 | - | 2 | 212 | ||