siRNA Details
| Gene Symbol: Prkcq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276721.1 | NP_001263650.1 | 18 | 2172 | ||
| GenBank mRNA | AB020614.1 | - | 4 | 419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkcq| Gene Symbol: Prkcq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276721.1 | NP_001263650.1 | 18 | 2172 | ||
| GenBank mRNA | AB020614.1 | - | 4 | 419 | ||