siRNA Details
Gene Symbol: Herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053523.1 | NP_445975.1 | 6 | 876 | ||
XM_006255133.2 | XP_006255195.1 | 6 | 1189 | |||
GenBank mRNA | AB033771.1 | - | 6 | 876 | ||
BC061849.1 | - | 6 | 898 | 5621877 |
