siRNA Details
| Gene Symbol: Tpra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053534.2 | NP_445986.2 | 9, 10 | 878 | ||
| XM_006236866.2 | XP_006236928.1 | 8, 9 | 2117 | |||
| XM_006236868.2 | XP_006236930.1 | 8, 9 | 918 | |||
| GenBank mRNA | AF292116.1 | - | 7, 8 | 684 | ||
| FQ228747.1 | - | 9, 10 | 953 | |||

Tpra1
Genomic Map