siRNA Details
| Gene Symbol: Cx3cl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134455.1 | NP_604450.1 | 3 | 939 | ||
| XM_008772329.1 | XP_008770551.1 | 4 | 1676 | |||
| GenBank mRNA | AF030358.2 | - | 3 | 939 | ||
| BC070938.1 | - | 3 | 940 | 7099250 | ||
| Y16813.1 | - | 1 | 310 | |||

Cx3cl1
Genomic Map