siRNA Details
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053550.1 | NP_446002.1 | 1 | 206 | ||
GenBank mRNA | AF033355.1 | - | 1 | 206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053550.1 | NP_446002.1 | 1 | 206 | ||
GenBank mRNA | AF033355.1 | - | 1 | 206 |