siRNA Details
| Gene Symbol: Pip4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053550.1 | NP_446002.1 | 3 | 391 | ||
| XM_008768131.1 | XP_008766353.1 | 3 | 670 | |||
| GenBank mRNA | AF033355.1 | - | 3 | 391 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pip4k2b| Gene Symbol: Pip4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053550.1 | NP_446002.1 | 3 | 391 | ||
| XM_008768131.1 | XP_008766353.1 | 3 | 670 | |||
| GenBank mRNA | AF033355.1 | - | 3 | 391 | ||