siRNA Details
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053550.1 | NP_446002.1 | 9 | 1203 | ||
XM_008768131.1 | XP_008766353.1 | 9 | 1482 | |||
GenBank mRNA | AF033355.1 | - | 9 | 1203 |
