siRNA Details
| Gene Symbol: Tnfsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053552.1 | NP_446004.1 | 3 | 650 | ||
| GenBank mRNA | AF037067.1 | - | 3 | 650 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfsf4| Gene Symbol: Tnfsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053552.1 | NP_446004.1 | 3 | 650 | ||
| GenBank mRNA | AF037067.1 | - | 3 | 650 | ||