siRNA Details
| Gene Symbol: Fez2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053600.2 | NP_446052.2 | 2 | 351 | ||
| XM_006239637.2 | XP_006239699.1 | 2 | 402 | |||
| XM_006239638.2 | XP_006239700.1 | 2 | 402 | |||
| XM_006239639.2 | XP_006239701.1 | 2 | 402 | |||
| GenBank mRNA | AB076183.1 | - | 2 | 351 | ||
| AB080086.1 | - | 2 | 330 | |||
| BC100060.1 | - | 2 | 332 | 7128839 | ||
| CK359698.1 | - | 2 | 318 | |||
| U64689.1 | - | 2 | 177 | |||

Fez2
Genomic Map