siRNA Details
| Gene Symbol: Pecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133299.2 | NP_579833.1 | 1 | 110 | ||
| XM_008767180.1 | XP_008765402.1 | 1 | 116 | |||
| GenBank mRNA | AF021854.1 | - | 1 | 111 | ||
| AF099742.1 | - | 1 | 30 | |||
| BC060546.1 | - | 1 | 104 | 6889831 | ||

Pecr
Genomic Map