siRNA Details
| Gene Symbol: Csn1s2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105741.1 | NP_001099211.1 | 9 | 348 | ||
| GenBank mRNA | J00712.1 | - | 9 | 348 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csn1s2a| Gene Symbol: Csn1s2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105741.1 | NP_001099211.1 | 9 | 348 | ||
| GenBank mRNA | J00712.1 | - | 9 | 348 | ||