siRNA Details
| Gene Symbol: Phb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013035.1 | NP_001013053.1 | 4, 5 | 608 | ||
| GenBank mRNA | BC083705.1 | - | 4, 5 | 608 | 7190294 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phb2| Gene Symbol: Phb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013035.1 | NP_001013053.1 | 4, 5 | 608 | ||
| GenBank mRNA | BC083705.1 | - | 4, 5 | 608 | 7190294 | |